Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Vinyl gloves, Rotiprotect®

Vinyl gloves, Rotiprotect®

Powdered, M, transparent, ≥240 mm, 0,18 mm, 100/box
Katalog # ROTH1904.1
Marketsource
Leverandør:   Roth Carl
Vinyl gloves, Rotiprotect®
Katalog # ROTH1904.1
Leverandør:  Roth Carl
Marketsource
Vinyl gloves, Rotiprotect®

Powdered, M, transparent, ≥240 mm, 0,18 mm, 100/box

Price
Spørg for pris
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents

Flexible, ambidextrous gloves made of PVC. Ideal for food handling.

  • Non sterile, powdered/powder-free, smooth finish
  • Very good sense of touch
  • With roll-type edge
  • Free of latex proteins

 : EN 455, Cat. I; AQL 1,5
Food contact approved. Not suitable for foodstuffs containing grease.

Specifikationer
  • Handske størrelse:
    M
  • Pakket:
    100/box
  • Farve:
    transparent
  • Længde:
    ≥240 mm
  • Seq:
    02
  • Tykkelse:
    0,18 mm
  • Model:
    Powdered
  • Cat. No.:
    ROTH1904.1
  • Ambidextrous/Hånd-specifik:
    Ambidextrous
  • Godkendt til fødevarekontakt:
    +
  • Med pudder/pudderfri:
    Med pudder
  • Latex/latex-fri:
    Latex-fri
  • Manchet:
    Rullet kant
  • EN norm:
    EN 455
  • Overflade:
    Smooth
  • Engangs/Genbrugelig:
    Engangs