Pierce™ Protein A
Protein A, 5 mg
Katalog # PIER21181
Leverandør: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Protein A can be used to probe and detect rabbit and human antibodies, especially IgG isotypes, in Western blotting, ELISA, IHC and other immunoassay protocols.
- Contains four Fc-binding domains per protein
- Efficient than Protein G for polyclonal IgG from rabbit, pig, dog and cat serum
- Less efficient than Protein G for mouse IgG1, human IgG3 and rat, goat and cow antibodies
Specifications
Specifications
- Pakn:5 mg
- Beskrivelse:Protein A