Orders LinkContactUsLinkComponent
Pierce™ Protein A
Pierce™ Protein A
Protein A, 5 mg
Katalog # PIER21181
Leverandør:   Thermo Fisher Scientific
Pierce™ Protein A
Katalog # PIER21181
CAS nummer:  
Pierce™ Protein A
Pierce™ Protein A
Protein A, 5 mg
CAS nummer:  
Price
1.990,00 kr
Log in for availability
1 * 5 mg
Product Details & Documents
Protein A can be used to probe and detect rabbit and human antibodies, especially IgG isotypes, in Western blotting, ELISA, IHC and other immunoassay protocols.
  • Contains four Fc-binding domains per protein
  • Efficient than Protein G for polyclonal IgG from rabbit, pig, dog and cat serum
  • Less efficient than Protein G for mouse IgG1, human IgG3 and rat, goat and cow antibodies
Specifications
Specifications
  • Pakn:
    5 mg
  • Beskrivelse:
    Protein A
Specifications