5,5'-Dithiobis(2-nitrobenzosyre)
250 Tests
Katalog # CAYM400050-250
Leverandør: Cayman Chemical
5,5'-Dithiobis(2-nitrobenzosyre)
Katalog # CAYM400050-250
Leverandør: Cayman Chemical
CAS nummer: 69-78-3
Gå til:
Product Details & Documents
Synonymer: DTNB, Ellman's reagent, 3,3'-Dithiobis(6-nitrobenzoic acid), 5,5'-Disulphanediylbis(2-nitrobenzoic acid), 2,2'-Dinitro-5,5'-dithiodibenzoic acid
- CAS nummer:69-78-3
- Formel:C₁₄H₈N₂O₈S₂
- MDL nummer:MFCD00007140
- Molecular Weight:396.36 g/mol
- Smeltende Pt:240…244 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-714-4
Specifikationer
Specifikationer
- Pakn:250 Tests



