Abbe refractometers, NAR
NAR-2T, 0 − 95 °Bx, 1,30 − 1,70 nD
Catalog # ATAG1220
Supplier: Atago
Jump to:
Product Details & Documents
The NAR Abbe refractometers can meet many of the requirements for routine measurements.
Single wavelengths and standard temperatures can be measured at extended ranges, as well as elevated temperatures, measurements of solid samples (films, plastic sheets).
Specifications
- Model:NAR-2T
- Område:0 − 95 °Bx
- Refraktionsindeks:1,30 − 1,70 nD
- Cat. No.:ATAG1220
- Lyskilde:LED'er
- Temperatursensorens nøjagtighed:±0,2/0,5 °C
- Temperature sensor:±0,2 °C
- Temperaturmåling:5...120 °C
Accessories
Product Information
- Catalog #BeskrivelseAvailabilityPrice
Product Information
- Catalog #BeskrivelseAvailabilityPrice
- ATAGRE-1165Termodisplayenhed (DTM-1), til NAR-1T•LO, NAR-2T•LO og NAR-2T•HILog in for availability1 * 1 Items
Product Information
- Catalog #BeskrivelseAvailabilityPrice
Product Information
- Catalog #BeskrivelseAvailabilityPrice