Natriumhydroxid ≥97%, pellets, Proteomics Grade
HDPE bottle, 1 kg
Leverandør: VWR Chemicals
Natriumhydroxid ≥97%, pellets, Proteomics Grade
Katalog # M137-1KG
Leverandør: VWR Chemicals
CAS nummer: 1310-73-2
Gå til:
Product Details & Documents
Synonymer: Caustic soda
- CAS nummer:1310-73-2
- Formel:NaOH
- MDL nummer:MFCD00003548
- Molecular Weight:40 g/mol
- Merck Index:14,08627
- Kogende Pt:1390 °C (1013 hPa)
- Smeltende Pt:318 °C
- Tæthed:2,13 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:215-185-5
- FN:1823
- ADR:8,II
- NÅ:01-2119457892-27
Documents
Specifikationer
Specifikationstestresultater
- Chloride:<0.005%
- DNase:none detected
- Heavy Metals:<0.002 %
- Iron:≤0.001%
- Mercury:≤0.1 ppm
- Nickel:≤0.001%
- Nitrogen Compounds:≤0.001%
- Phosphate:≤0.001%
- Potassium:≤0.02%
- Protease:none
- Purity:≥97%
- RNase:none detected
- Sodium Carbonate:≤1%
- Sulphate:≤0.003%
Specifikationer
- Pakning:HDPE bottle
- Pakn:1 kg



