2-Mercaptoethanol ≥99%, Proteomics Grade
Bottle, amber, 100 mL
Leverandør: VWR Chemicals
2-Mercaptoethanol ≥99%, Proteomics Grade
Katalog # M131-100ML
Leverandør: VWR Chemicals
CAS nummer: 60-24-2
Gå til:
Product Details & Documents
Synonymer: 2-Hydroxyethyl mercaptan, β-Mercaptoethanol
- CAS nummer:60-24-2
- Formel:HSCH₂CH₂OH
- MDL nummer:MFCD00004890
- Molecular Weight:78.14 g/mol
- Merck Index:13,05894
- Kogende Pt:157 °C (1013 hPa)
- Smeltende Pt:–100 °C
- Tæthed:1,12 g/cm³ (20 °C)
- Flash Pt:68 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-464-6
- FN:2966
- ADR:6.1,II
Documents
Specifikationer
Specifikationstestresultater
- DNase:none detected
- Protease:none detected
- Purity:≥99%
- RNase:none detected
- Water:≤0.5%
Specifikationer
- Pakning:Bottle, amber
- Pakn:100 mL



