Orders LinkContactUsLinkComponent
VWR®, Tris-borate-EDTA (TBE) buffer
VWR®, Tris-borate-EDTA (TBE) buffer
TBE buffer solution 5X concentrate (TRIS-borate-EDTA buffer), ultrapure, Bag-in-box, 4 L
Katalog # J885-4L
Leverandør:   VWR Chemicals
VWR®, Tris-borate-EDTA (TBE) buffer
Katalog # J885-4L
Leverandør:  VWR Chemicals
CAS nummer:  
VWR®, Tris-borate-EDTA (TBE) buffer
VWR®, Tris-borate-EDTA (TBE) buffer
TBE buffer solution 5X concentrate (TRIS-borate-EDTA buffer), ultrapure, Bag-in-box, 4 L
CAS nummer:  
Price
1.600,00 kr
Log ind for tilgængelighed
1 * 4 L
Product Details & Documents
TBE is optimised for DNA electrophoresis techniques using both acrylamide and agarose. Buffers are available as liquid concentrate or Ready-Pack™ (a foil pouch containing sufficient material to prepare 1 litre of 10X concentrate).
  • High buffering capacity
  • High ionic strength
  • Allows extended electrophoresis runs without buffer recirculation
  • Convenient packaging options minimise weighing and mixing
  • Free from DNase, RNase and protease
Composition of (1X) solution contains 0,089 M Tris base, 0,089 M borate and 0,002 M EDTA, disodium salt, dihydrate with a final pH of 8,3.
Caution: *Note: TBE has a tendency to precipitate over time. If this occurs, redissolve in solution.
Specifikationer
Specifikationer
  • Pakning:
    Bag-in-box
  • Pakn:
    4 L
  • Beskrivelse:
    TBE buffer solution 5X concentrate (TRIS-borate-EDTA buffer), ultrapure