Orders LinkContactUsLinkComponent
VWR®, Yeast extract
VWR®, Yeast extract
Yeast extract, 1 kg
Katalog # J850-1KG
Leverandør:   VWR Chemicals
VWR®, Yeast extract
Katalog # J850-1KG
Leverandør:  VWR Chemicals
CAS nummer:  
VWR®, Yeast extract
VWR®, Yeast extract
Yeast extract, 1 kg
CAS nummer:  
Price
2.200,00 kr
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents
Yeast extract is the water soluble portion of autolysed yeast containing a source of Vitamin B complex.
  • High purity extract
  • Used for supplementing media
Documents
Specifikationer
Specifikationer
  • Pakn:
    1 kg
  • Beskrivelse:
    Yeast extract