Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Natriumiodid, Ultrapure

Natriumiodid, Ultrapure

HDPE bottle, 500 g
Katalog # 0853-500G
Miljø
Leverandør:   VWR Chemicals
Natriumiodid, Ultrapure
Katalog # 0853-500G
Leverandør:  VWR Chemicals
CAS nummer:  7681-82-5
Natriumiodid, Ultrapure
Natriumiodid, Ultrapure

HDPE bottle, 500 g

CAS nummer:  7681-82-5
Price
3.360,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents
Synonymer:  Hydriodic acid sodium salt, Sodium monoiodide, Ioduril
  • CAS nummer:7681-82-5
  • Formel:NaI
  • MDL nummer:MFCD00003532
  • Molecular Weight:149.89 g/mol
  • Merck Index:13,08705
  • Kogende Pt:1300 °C (1013 hPa)
  • Smeltende Pt:662 °C
  • Tæthed:3,67 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:231-679-3
  • FN:3077
  • ADR:9,III
Specifikationer

Specifikationer

  • Pakning:
    HDPE bottle
  • Pakn:
    500 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.