Natriumiodid, Ultrapure
HDPE bottle, 250 g
Leverandør: VWR Chemicals
Gå til:
Product Details & Documents
Synonymer: Hydriodic acid sodium salt, Sodium monoiodide, Ioduril
- CAS nummer:7681-82-5
- Formel:NaI
- MDL nummer:MFCD00003532
- Molecular Weight:149.89 g/mol
- Merck Index:13,08705
- Kogende Pt:1300 °C (1013 hPa)
- Smeltende Pt:662 °C
- Tæthed:3,67 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:231-679-3
- FN:3077
- ADR:9,III
Documents
Specifikationer
Specifikationer
- Pakning:HDPE bottle
- Pakn:250 g



