Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Polysorbat 20, Reagent Grade

Cube container, 4 L
Catalog # 0777-4L
Supplier:   VWR Chemicals
Polysorbat 20, Reagent Grade
Catalog # 0777-4L
Supplier:  VWR Chemicals
CAS Number:  9005-64-5
Polysorbat 20, Reagent Grade
Polysorbat 20, Reagent Grade

Cube container, 4 L

CAS Number:  9005-64-5
Price
1.390,00 kr
Log in for availability
1 * 4 L
Product Details & Documents

Non-ionic detergent for solubilising membrane proteins without altering biological activity.

Synonyms:  Sorbitanmonolaurat, ethoxyleret
  • CAS Number:9005-64-5
  • Formula:C₅₈H₁₁₄O₂₆
  • MDL Number:MFCD00165986
  • Boiling Pt:>100 °C
  • Density:1,1 g/cm³ (20 °C)
  • Flash Pt:149 °C (closed cup)
  • Storage Temperature:Stue temperatur
  • EINECS:500-018-3
Specifications

Specification Test Results

  • Arsenic:
    <0.0003%
  • DNase:
    NONE
  • Heavy Metals:
    <0.001%
  • Hydroxyl Number:
    96 – 108
  • Identification:
    PASS
  • Moisture (KF):
    3.0%
  • Protease:
    NONE
  • Residue on Ignition:
    0.25%
  • RNase:
    NONE

Specifications

  • Pakning:
    Cube container
  • Pakn:
    4 L

Specifications