Orders LinkContactUsLinkComponent
VWR®, Agarose I™
VWR®, Agarose I™
Agarose I™
Katalog # 0710-500G
Leverandør:   VWR Chemicals
VWR®, Agarose I™
Katalog # 0710-500G
Leverandør:  VWR Chemicals
VWR®, Agarose I™
VWR®, Agarose I™
Agarose I™
Price
7.190,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents
Agarose I™ is a standard melting/gelling agarose, suitable for routine nucleic acid analytical/preparative applications (250 bp to 22 kb).
  • All-purpose, high purity agarose
  • Exceptional band resolution and clarity
  • Nuclease- and protease-free
Agarose I™ has a low EEO (electroendosmosis) for shorter electrophoretic runs without compromising resolution. Agarose I™ is excellent for blotting techniques and general manipulations.
Specifikationer
  • Beskrivelse:
    Agarose I™
  • Seq:
    04