VWR®, Agarose I™
Agarose I™
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Agarose I™ is a standard melting/gelling agarose, suitable for routine nucleic acid analytical/preparative applications (250 bp to 22 kb).
- All-purpose, high purity agarose
- Exceptional band resolution and clarity
- Nuclease- and protease-free
Agarose I™ has a low EEO (electroendosmosis) for shorter electrophoretic runs without compromising resolution. Agarose I™ is excellent for blotting techniques and general manipulations.
Documents
Specifications
- Beskrivelse:Agarose I™
- Seq:01