Orders LinkContactUsLinkComponent
VWR®, Agarose I™
VWR®, Agarose I™
Agarose I™
Catalog # 0710-25G
Supplier:   VWR Chemicals
VWR®, Agarose I™
Catalog # 0710-25G
Supplier:  VWR Chemicals
VWR®, Agarose I™
VWR®, Agarose I™
Agarose I™
Price
810,00 kr
Log in for availability
1 * 25 g
Product Details & Documents
Agarose I™ is a standard melting/gelling agarose, suitable for routine nucleic acid analytical/preparative applications (250 bp to 22 kb).
  • All-purpose, high purity agarose
  • Exceptional band resolution and clarity
  • Nuclease- and protease-free
Agarose I™ has a low EEO (electroendosmosis) for shorter electrophoretic runs without compromising resolution. Agarose I™ is excellent for blotting techniques and general manipulations.
Specifications
  • Beskrivelse:
    Agarose I™
  • Seq:
    01