Orders LinkContactUsLinkComponent
VWR®, Tris-borate-EDTA (TBE) buffer
TBE buffer, 10X liquid concentrate, Bag-in-box, 4 L
Catalog # 0658-4L
Supplier:   VWR Chemicals
VWR®, Tris-borate-EDTA (TBE) buffer
Catalog # 0658-4L
Supplier:  VWR Chemicals
CAS Number:  
VWR®, Tris-borate-EDTA (TBE) buffer
VWR®, Tris-borate-EDTA (TBE) buffer
TBE buffer, 10X liquid concentrate, Bag-in-box, 4 L
CAS Number:  
Price
1.030,00 kr
Log in for availability
1 * 4 L
Product Details & Documents
TBE is optimised for DNA electrophoresis techniques using both acrylamide and agarose. Buffers are available as liquid concentrate or Ready-Pack™ (a foil pouch containing sufficient material to prepare 1 litre of 10X concentrate).
  • High buffering capacity
  • High ionic strength
  • Allows extended electrophoresis runs without buffer recirculation
  • Convenient packaging options minimise weighing and mixing
  • Free from DNase, RNase and protease
Composition of (1X) solution contains 0,089 M Tris base, 0,089 M borate and 0,002 M EDTA, disodium salt, dihydrate with a final pH of 8,3.
 : *Note: TBE has a tendency to precipitate over time. If this occurs, redissolve in solution.
Specifications
Specifications
  • Pakning:
    Bag-in-box
  • Pakn:
    4 L
  • Beskrivelse:
    TBE buffer, 10X liquid concentrate
Specifications