Natriumdeoxycholat ≥99.0%, high purity
HDPE bottle, 100 g
Leverandør: VWR Chemicals
Natriumdeoxycholat ≥99.0%, high purity
Katalog # 0613-100G
Leverandør: VWR Chemicals
CAS nummer: 302-95-4
Gå til:
Product Details & Documents
Anionic - negatively-charged (carboxylates, sulphates or sulphonates in alkali metal form). Utilised for applications ranging from solubilising membrane proteins in non-denaturing conditions to creating denaturing conditions during gel electrophoresis. Used for complete disruption of membranes.
Synonymer: Deoxycholic acid sodium salt, 3α,12α-Dihydroxy-5β-cholan-24-oic acid sodium salt, Sodium 3α,12α-dihydroxy-5β-cholanoate
- CAS nummer:302-95-4
- Formel:C₂₄H₃₉NaO₄
- MDL nummer:MFCD00064139
- Molecular Weight:414.56 g/mol
- Smeltende Pt:357…365 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:206-132-7
Documents
Specifikationer
Specifikationstestresultater
- Arsenic:≤0.001%
- Heavy metals:≤0.005%
- Loss on drying:≤5.0%
- Purity (dry basis):≥99.0%
- Sodium cholate:≤2.0%
- Solubility (10%, water):Pass
Specifikationer
- Pakning:HDPE bottle
- Pakn:100 g



