Natriumhydroxid ≥97%, pellets ACS
HDPE bottle, 1 kg
: Caustic soda
- :1310-73-2
- :NaOH
- :MFCD00003548
- :40 g/mol
- :14,08627
- :1390 °C (1013 hPa)
- :323 °C
- :2,13 g/cm³ (20 °C)
- :Stue temperatur
- :215-185-5
- :1823
- :8,II
- :01-2119457892-27
- Calcium:<0.005%
- Chloride:≤0.005%
- Heavy metals:≤0.002%
- Iron:≤0.001%
- Magnesium:≤0.002%
- Mercury:≤0.1 ppm
- Nickel:≤0.001%
- Nitrogen compounds:≤0.001%
- Phosphate:≤0.001%
- Potassium:≤0.02%
- Purity:≥97%
- Sodium carbonate:≤1%
- Sulphate:≤0.003%
- Pakning:HDPE bottle
- Pakn:1 kg



