Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Natriumpyruvat ≥99%, Reagent Grade

Natriumpyruvat ≥99%, Reagent Grade

Plastic bottle for solids, 250 g
Katalog # 0342-250G
Leverandør:   VWR Chemicals
Natriumpyruvat ≥99%, Reagent Grade
Katalog # 0342-250G
Leverandør:  VWR Chemicals
CAS nummer:  113-24-6
Natriumpyruvat ≥99%, Reagent Grade
Natriumpyruvat ≥99%, Reagent Grade

Plastic bottle for solids, 250 g

CAS nummer:  113-24-6
Price
2.650,00 kr
Log ind for tilgængelighed
1 * 250 g
Product Details & Documents
Synonymer:  2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt
  • CAS nummer:113-24-6
  • Formel:CH₃COCOONa
  • MDL nummer:MFCD00002586
  • Molecular Weight:110.04 g/mol
  • Smeltende Pt:220…230 °C
  • Tæthed:1,718 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:204-024-4
Specifikationer

Specifikationstestresultater

  • Free Acid:
    ≤1
  • Heavy Metals (as Pb):
    ≤0.01%
  • Water Content:
    <1%
  • Purity:
    ≥99%

Specifikationer

  • Pakning:
    Plastic bottle for solids
  • Pakn:
    250 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.