Natriumpyruvat ≥99%, Reagent Grade
Plastic bottle for solids, 100 g
: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt
- :113-24-6
- :CH₃COCOONa
- :MFCD00002586
- :110.04 g/mol
- :220…230 °C
- :1,718 g/cm³ (20 °C)
- :Stue temperatur
- :204-024-4
- Free Acid:≤1
- Heavy Metals (as Pb):≤0.01%
- Water Content:<1%
- Purity:≥99%
- Pakning:Plastic bottle for solids
- Pakn:100 g
- Cat. No.:0342-100G



