Natriumhydroxid, beads, Reagent Grade
HDPE bottle, 1 kg
Supplier: VWR Chemicals
Natriumhydroxid, beads, Reagent Grade
Catalog # 0173-1KG
Supplier: VWR Chemicals
CAS Number: 1310-73-2
Jump to:
Product Details & Documents
Synonyms: Caustic soda
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- Merck Index:14,08627
- Boiling Pt:1390 °C (1013 hPa)
- Melting Pt:323 °C
- Density:2,13 g/cm³ (20 °C)
- Storage Temperature:Stue temperatur
- EINECS:215-185-5
- UN:1823
- ADR:8,II
- REACH:01-2119457892-27
Documents
Certificates
Search by Lot NumberSpecifications
Specification Test Results
- Chloride:≤0.05%
- Heavy Metals:≤0.006%
- Potassium:≤0.1%
- Purity:>98.5%
- Sodium Carbonate:≤0.90%
- Sulphate:≤0.005%
Specifications
- Pakning:HDPE bottle
- Pakn:1 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



