Capillaries for LC systems
Flexible capillaries, green, stainless steel, without fittings, 4,6 μl, 200 mm, I.Ø 0,17 mm
Catalog # HEWL5065-9931
Supplier: AGILENT HEWLETT PACKARD
Jump to:
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifications
- Beskrivelse:Flexible capillaries, green, stainless steel, without fittings, 4,6 μl, 200 mm, I.Ø 0,17 mm
- Seq:101