Orders LinkContactUsLinkComponent
Racks for freezer
Racks for freezer
12 (3×4), 50 mm, 424×140×219 mm
Katalog # RATI5400009
Leverandør:   RATIOLAB
Racks for freezer
Katalog # RATI5400009
Leverandør:  RATIOLAB
Racks for freezer
12 (3×4), 50 mm, 424×140×219 mm
Price
1.160,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
For the storing of cryoboxes in freezer cabinets and chests.
  • Robust construction
  • Handles for convenient transport
  • Made of stainless steel with surfaces for perfect cleaning
Leveringsinformation: Cryo racks are supplied with shelf sizes for any standard cryoboxes.
Specifikationer
  • Rum:
    12 (3×4)
  • B×D×H udv.:
    424×140×219 mm
  • Seq:
    01
  • Cryobox højde:
    50 mm