Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Filter Holder Accessories

Tubing, 6 mm, O.Ø 3 m, polypropylene
Katalog # MILFXX6702508
Leverandør:   Merck
Filter Holder Accessories
Katalog # MILFXX6702508
Leverandør:  Merck
Filter Holder Accessories
Filter Holder Accessories

Tubing, 6 mm, O.Ø 3 m, polypropylene

Price
661,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Tubing, 6 mm, O.Ø 3 m, polypropylene
  • Seq:
    06
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.