Stirring shafts
WB 10, 240 mm, 10 mm
Katalog # LENZ06300510
Leverandør: Lenz Laborglas
Gå til:
Product Details & Documents
Stirring shafts made of precision calibrated glass tube.
- Ground and polished bearing surface
- Interchangeable
- Made of DURAN® tubing
Documents
BSE TSE Glass Columns
BSE/TSE-certifikater
Specifikationer
- Ø:10 mm
- Model:WB 10
- Længde:240 mm
- Seq:01