Methylacetat, Supelco®
Glass bottle, 5 mL
Katalog # 45997-5ML-F
Leverandør: Merck
Gå til:
Product Details & Documents
Synonymer: Acetic acid methyl ester
- CAS nummer:79-20-9
- Formel:C₃H₆O₂
- MDL nummer:MFCD00008711
- Molecular Weight:74.08 g/mol
- Merck Index:13,06038
- Kogende Pt:56,9 °C (1013 hPa)
- Smeltende Pt:–98 °C
- Tæthed:0,938 g/cm³ (20 °C)
- Flash Pt:–13 °C
- Opbevaringstemperatur:Stue temperatur
- FN:1231
- ADR:3,II
- :M-Clarity™ quality level = MQ100
- EINECS:201-185-2
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:5 mL



