Anisol, Supelco®
Glass bottle, 5 mL
Katalog # 96109-5ML-F
Leverandør: Merck
Gå til:
Product Details & Documents
Synonymer: Methyl phenyl ether, Methoxybenzene
- CAS nummer:100-66-3
- Formel:C₇H₈O
- MDL nummer:MFCD00008354
- Molecular Weight:108.14 g/mol
- Merck Index:13,00672
- Kogende Pt:154 °C (1013 hPa)
- Smeltende Pt:–37 °C
- Tæthed:0,9961 g/cm³ (20 °C)
- Flash Pt:44 °C
- Opbevaringstemperatur:Stue temperatur
- FN:2222
- ADR:3,III
- :M-Clarity™ quality level = MQ100
- EINECS:202-876-1
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:5 mL
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



