Orders LinkContactUsLinkComponent
Instrument trays
Instrument trays
150×190×17 mm
Katalog # BURK4230-3019
Leverandør:   BURKLE
Instrument trays
Katalog # BURK4230-3019
Leverandør:  BURKLE
Instrument trays
Instrument trays
150×190×17 mm
Price
96,70 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Melamine, white.
  • Tray without lid
    Suitable for use with foodstuffs
    Not autoclavable
    Not suitable for high temperatures
Ordering information: Lid available as accessory
Specifikationer
  • B×D×H:
    150×190×17 mm