Orders LinkContactUsLinkComponent
Dressing forceps, simple type
Titanium, curved, blunt end tips, 200 mm
Katalog # HAMMHTC081-20
Leverandør:   HAMMACHER KARL
Dressing forceps, simple type
Katalog # HAMMHTC081-20
Leverandør:  HAMMACHER KARL
Dressing forceps, simple type
Dressing forceps, simple type
Titanium, curved, blunt end tips, 200 mm
Price
739,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Titanium.
Specifikationer
  • Model:
    Titanium, curved, blunt end tips
  • Længde:
    200 mm
  • Seq:
    02