Orders LinkContactUsLinkComponent
Dråbeflasker, WHEATON®
Dråbeflasker, WHEATON®
20-410, orange
Katalog # 215-4045
Leverandør:   DWK Life Sciences
Dråbeflasker, WHEATON®
Katalog # 215-4045
Leverandør:  DWK Life Sciences
Dråbeflasker, WHEATON®
Dråbeflasker, WHEATON®
20-410, orange
Price
4.870,00 kr
Log ind for tilgængelighed
1 * 1.000 Items
Product Details & Documents
Screw cap made of radiation grade polypropylene.
  • Very good chemical resistance
  • Very good temperature resistance
  • Variety of colors
Specifikationer
  • Gevind:
    20-410
  • Seq:
    48
  • Lågfarve:
    orange