Scintillation vials, WHEATON®
20,00 ml, 27×61 mm, Yes, Separate, 22 mm, PP, Metal foil
Katalog # WHEA986711
Leverandør: DWK Life Sciences
Gå til:
Product Details & Documents
These scintillation vials made from HDPE are available with screw cap attached or separately in PP or PE and different liner material depending on the application.
Specifikationer
- Kasket inkluderet:Yes
- Hættemateriale:PP
- Cap størrelse:22 mm
- Diameter×H:27×61 mm
- Septum:Metal foil
- Kapacitet:20,00 ml
- Seq:08
- Låg type:Separate