Drum thieves, Wheaton®
Drum thieves, Glas, 150 ml
WHEA885303
Drum thieves made of soda lime glass for quick disposal.
- Conventional open end drum thieves
- Just insert empty tube in drum
- Hold thumb over top and withdraw
- Kapacitet:150 ml
- Beskrivelse:Drum thieves
- Materiale:Glas