Orders LinkContactUsLinkComponent
Arsine generator apparatus, modified, WHEATON®
Vaskeabsorberrør
Katalog # WHEA180024
Leverandør:   DWK Life Sciences
Arsine generator apparatus, modified, WHEATON®
Katalog # WHEA180024
Leverandør:  DWK Life Sciences
Arsine generator apparatus, modified, WHEATON®
Vaskeabsorberrør
Price
1.600,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Modified version for arsine analysis which eliminates the cumbersome ball and socket joint connecting the scrubber and absorber tubes which conforms to ASTM / USP and EPA specifications.
The freestanding system has a standard taper 24/40 Clear-Seal™ joint. Manufactured from 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements.
Specifikationer
  • Beskrivelse:
    Vaskeabsorberrør
  • Seq:
    02