BOD bottles, WHEATON®
BOD bottles, with bar codes, borosilicate glass, 69×165 mm (Ø×H), number sequence: 289-312, 300 ml
Katalog # WHEA227497-13G
Leverandør: DWK Life Sciences
Gå til:
Product Details & Documents
Made of Type I borosilicate glass.
- Available with or without stopper
- With white labeling area
Specifikationer
- Kapacitet:300 ml
- Beskrivelse:BOD bottles, with bar codes, borosilicate glass, 69×165 mm (Ø×H), number sequence: 289-312
- Cat. No.:WHEA227497-13G