Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Stoppers for BOD bottles, WHEATON®

Stoppers for BOD bottles, WHEATON®

Pennyhead stopper
Katalog # WHEA227672
Leverandør:   DWK Life Sciences
Stoppers for BOD bottles, WHEATON®
Katalog # WHEA227672
Leverandør:  DWK Life Sciences
Stoppers for BOD bottles, WHEATON®
Stoppers for BOD bottles, WHEATON®

Pennyhead stopper

Price
1.020,00 kr
Log ind for tilgængelighed
1 * 12 Items
Product Details & Documents

Borosilicate glass.

  • Very good chemical resistance
  • Very good temperature resistance

For 60 ml and 300 ml BOD bottles.

Specifikationer
  • Beskrivelse:
    Pennyhead stopper
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.