Dråbeflasker, WHEATON®
20-410, lilla
Katalog # 215-4025
Leverandør: DWK Life Sciences
Gå til:
Product Details & Documents
Screw cap made of radiation grade polypropylene.
- Very good chemical resistance
- Very good temperature resistance
- Variety of colors
Specifikationer
- Gevind:20-410
- Seq:43
- Lågfarve:lilla