Engangs-baseforme
Deep base mould, for M512 MACROSETTE®, 37×24×10 mm
Catalog # 720-0208
Supplier: Simport Scientific
Jump to:
Product Details & Documents
These disposable PVC base moulds offer ease and convenience of operation. They can be discarded after use, but are also strong enough to be reused.
- Excellent thermal exchange
- Smooth interior
- Rounded corners
They are available in the same variety of sizes as metal moulds and can be used with the same styles or types of cassettes and embedding rings.
Delivery information: Each case contains two dispenser boxes of 500 base moulds.
Documents
Specifications
- Beskrivelse:Deep base mould, for M512 MACROSETTE®
- Dimensioner:37×24×10 mm
- Seq:06