Eddikesyre glacial ≥98%, TECHNICAL
Plastic bottle, 1 L
Leverandør: VWR Chemicals
Gå til:
Product Details & Documents
Synonymer: Vinegar acid, Methanecarboxylic acid, Ethanoic acid
- CAS nummer:64-19-7
- Formel:H₃CCOOH
- MDL nummer:MFCD00036152
- Molecular Weight:60.05 g/mol
- Kogende Pt:118 °C (1013 hPa)
- Smeltende Pt:17 °C
- Tæthed:1,05 g/cm³ (20 °C)
- Flash Pt:38,5 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-580-7
- FN:2789
- ADR:8 (3),II
- NÅ:01-2119475328-30
Documents
Certifikater
Søg efter partinummer1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Emballageoplysninger
P20_VWRC84528.290_SPEC_EN.pdf P20_VWRC84528.360_SPEC_EN.pdf P20_VWRC84528.460_SPEC_EN.pdf spec_VWRC84528.290 spec_VWRC84528.290
Produktspecifikation
More
VWR Chemicals Big Packs and Bulks
Produktlitteratur
Specifikationer
Specifikationstestresultater
- Assay:Min. 98 %
Specifikationer
- Pakning:Plastic bottle
- Pakn:1 L
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



