Trimming knives
SM 53
Katalog # SWAN4206
Leverandør: SWANN MORTON
Gå til:
Product Details & Documents
Heavy duty trimming knifes with fixed or retractable blade.
- Blades can be exchanged
- Trimming knife with a smart black holster that clips onto a belt or waistband
Leveringsinformation: The fixed trimming knife is supplied with holster and three SM92 industrial blades. The retractable knife is supplied with two SM92 industrial blades.
Specifikationer
- Model:SM 53