Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Craft knife blades

Craft knife blades

Nr. 15
Katalog # SWAN9275
Leverandør:   SWANN MORTON
Craft knife blades
Katalog # SWAN9275
Leverandør:  SWANN MORTON
Craft knife blades
Craft knife blades

Nr. 15

Price
574,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents

Surgical quality blades for use with handles No. 5A and No. 6A.

Packaging: Supplied in anti-corrosion packaging.

Specifikationer
  • Model:
    Nr. 15
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.