Orders LinkContactUsLinkComponent
Stitch cutters
Stitch cutters
Standardsuturskærer, afrundet spids, kulstofstål, sterilt
Katalog # SWAN0420
Leverandør:   SWANN MORTON
Stitch cutters
Katalog # SWAN0420
Leverandør:  SWANN MORTON
Stitch cutters
Stitch cutters
Standardsuturskærer, afrundet spids, kulstofstål, sterilt
Price
448,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
These sterile stitch cutters are made from either carbon steel or stainless steel.
  • Convenient and cost effective
  • For use without a handle
The traditional carbon steel standard stitch cutter with its rounded bull nose tip which ensures there is no damage to the skin whilst the surgical edge cuts through the suture with minimal resistance.
The long stitch cutter allows for a more aseptic techinique by operating further away from the actual wound which also permits better visibility
Specifikationer
  • Beskrivelse:
    Standardsuturskærer, afrundet spids, kulstofstål, sterilt