Post-mortem scalpel handles
PM40H bright zinc plated, Blade no's PM40 and PM40B, 220 mm
Leverandør: SWANN MORTON
Gå til:
Product Details & Documents
A versatile and comprehensive range for morticians and pathologists for use in autopsy, histopathology and general post-mortem procedures.
- PM40 range with traditional screw fitting
- PM60 range with standard blade fitting
PM40B and PM60B blades are 'bull nose' blades for added safety and protection against accidental sharps injuries.
: The PM8 and PM40 stainless steel handles and all blades are supplied individually wrapped.
Specifikationer
- Til:Blade no's PM40 and PM40B
- Model:PM40H bright zinc plated
- Længde:220 mm
- Seq:04