Orders LinkContactUsLinkComponent
Membrane filters with grid
Membrane filters with grid
White with black grid, cellulose nitrate, 6 mm hydrophobic edge, 131, 0,45 µm, 47 mm
Katalog # 611-1157
Leverandør:   Sartorius
Membrane filters with grid
Katalog # 611-1157
Leverandør:  Sartorius
Membrane filters with grid
Membrane filters with grid
White with black grid, cellulose nitrate, 6 mm hydrophobic edge, 131, 0,45 µm, 47 mm
Price
2.690,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
Hydrophobic cellulose nitrate and cellulose acetate gridded membrane filters for colony count and sterility testing, with outstanding retention rates for microorganisms.
Membranes with 0,45 µm pore size are tested and released according to ISO 7704.
Documents
Specifikationer
  • Ø:
    47 mm
  • Porestørrelse:
    0,45 µm
  • Model:
    131
  • Beskrivelse:
    White with black grid, cellulose nitrate, 6 mm hydrophobic edge
  • Seq:
    06
  • Tykkelse:
    115 - 145 µm
  • Cat. No.:
    611-1157
  • Max. temperature:
    130 °C