Microtitre plates, 96-well
Flad bund, PS, klar, 96, 300 µl
Katalog # 735-0194
Leverandør: Thermo Fisher Scientific
Microtitre plates, 96-well
Katalog # 735-0194
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Rigid styrene or flexible vinyl, square well. Ideal for micro-volume applications, such as storage, sample or reagent transfer, serial dilution, haemogglutination, blood typing and compliment fixation. Available with a choice of flat, U- or V-bottom well shape.
- U-bottom well shape improves washing in ELISAs, enhances sensitivity in fluorescent applications and facilitates observation of agglutination reactions
- V-bottom well shape is optimal for magnetic bead assays, ideal for agglutination tests, useful for centrifugation or concentration and facilitates recovery of contents
Specifikationer
- Ant. brønde:96
- Farve:klar
- Beskrivelse:Flad bund, PS
- Total volumen:300 µl
- Cat. No.:735-0194
- Miljøcertificering:NAR Certificate of Retirement
- Environmentally Preferable:Lav fremstillingspåvirkning