Orders LinkContactUsLinkComponent
Spatulas, knob end
Spatulas, knob end
210 mm, 70 mm, 11 mm
Catalog # 231-0046
Supplier:   BOCHEM
Spatulas, knob end
Catalog # 231-0046
Supplier:  BOCHEM
Spatulas, knob end
Spatulas, knob end
210 mm, 70 mm, 11 mm
Price
136,00 kr
Log in for availability
1 * 5 Items
Product Details & Documents
Stainless steel. One sided spatulas, available individually or in economical packs of five.
  • Heat and chemical resistant
Specifications
  • Længde:
    210 mm
  • Bladbredde:
    11 mm
  • Klinge længde:
    70 mm