Manikins
Manikin Geriatric Model Ii
Katalog # BINH1005597
Marketsource
Leverandør: 3B Scientific
Gå til:
Product Details & Documents
This full-size manikin allows the training of BLS rescue techniques and CPR. The resiliency and weight of the body is very realistic.
- Several models
Specifikationer
- Beskrivelse:Manikin Geriatric Model Ii