Orders LinkContactUsLinkComponent
Kryorør, Nunc™ CryoTubes™
Kryorør, Nunc™ CryoTubes™
Cryogenic tubes with writing area, 1,00 ml, V, 12,5×41 *** mm, Yes, klar
Katalog # 479-6846
Leverandør:   Thermo Fisher Scientific
Kryorør, Nunc™ CryoTubes™
Katalog # 479-6846
Kryorør, Nunc™ CryoTubes™
Kryorør, Nunc™ CryoTubes™
Cryogenic tubes with writing area, 1,00 ml, V, 12,5×41 *** mm, Yes, klar
Price
1.690,00 kr
Udsalg
Gælder indtil kl 30/06/2026
Log ind for tilgængelighed
1 * 500 Items
Product Details & Documents
PP tubes with PP screw cap (internal thread vials) or PE screw cap (external thread vials) and white writing area, sterile. Developed for storage of cell cultures, bacteria and other samples at extremely low temperatures. Suitable for use in freezers as well as in the vapour phase of liquid nitrogen*.
Non toxic, non pyrogenic.
Conform to IATA regulations for the air transportation of biological samples.
* Advarsel: Brug ikke CryoTubes™ i flydende nitrogens væskefase, med mindre de er korrekt forseglede i Nunc CryoFlex™-slanger.
* uden skrivefelt
** fritstående
*** stjernefod
Specifikationer
  • Kasket inkluderet:
    Yes
  • Diameter×H:
    12,5×41 *** mm
  • Kapacitet:
    1,00 ml
  • Beskrivelse:
    Cryogenic tubes with writing area
  • Seq:
    15
  • Lågfarve:
    klar
  • Grundlag:
    V