Identification labels holder, Aesculap®
Barcode label holder, with lettering, 50×35×13 mm
Katalog # AESCJG730B
Leverandør: Aesculap
Gå til:
Product Details & Documents
The holder for identification labels is used for mounting on trays.
- External dimensions: 50×35×13 mm
The holder with lettering, have maximum 14 characters per line and maximum character height is 4 mm.
Specifikationer
- Beskrivelse:Barcode label holder, with lettering
- Dimensioner:50×35×13 mm
- Seq:01