Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Storage tray for endoscopes, Aesculap®

Storage tray for endoscopes, Aesculap®

Double scope tray with silicone support, sheath length 340 mm, 454×104×41 mm
Katalog # AESCJF433R
Leverandør:   Aesculap
Storage tray for endoscopes, Aesculap®
Katalog # AESCJF433R
Leverandør:  Aesculap
Storage tray for endoscopes, Aesculap®

Double scope tray with silicone support, sheath length 340 mm, 454×104×41 mm

Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Double scope tray with silicone support, sheath length 340 mm
  • Seq:
    05
  • B×D×H:
    454×104×41 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.