Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Storage tray for endoscopes, Aesculap®

Storage tray for endoscopes, Aesculap®

Double scope tray, sheath length 340 mm, 454×104×41 mm
Catalog # AESCJF432R
Supplier:   Aesculap
Storage tray for endoscopes, Aesculap®
Catalog # AESCJF432R
Supplier:  Aesculap
Storage tray for endoscopes, Aesculap®

Double scope tray, sheath length 340 mm, 454×104×41 mm

Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Beskrivelse:
    Double scope tray, sheath length 340 mm
  • Seq:
    04
  • B×D×H:
    454×104×41 mm