Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Storage tray for endoscopes, Aesculap®

Single scope tray, sheath length 340 mm, 454×84×41 mm
Katalog # AESCJF431R
Leverandør:   Aesculap
Storage tray for endoscopes, Aesculap®
Katalog # AESCJF431R
Leverandør:  Aesculap
Storage tray for endoscopes, Aesculap®

Single scope tray, sheath length 340 mm, 454×84×41 mm

Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Single scope tray, sheath length 340 mm
  • Seq:
    03
  • B×D×H:
    454×84×41 mm
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.