Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Chisels, Aesculap®

Chisels, Aesculap®

Chisel, Stille, 205 mm, 20 mm
Katalog # AESCFL506R
Leverandør:   Aesculap
Chisels, Aesculap®
Katalog # AESCFL506R
Leverandør:  Aesculap
Chisels, Aesculap®

Chisel, Stille, 205 mm, 20 mm

Price
1.150,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

These chisels are used for dental surgery.

Specifikationer
  • Beskrivelse:
    Chisel
  • Længde:
    205 mm
  • Seq:
    17
  • Model:
    Stille
  • Bredde:
    20 mm
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.